Novus Biologicals
Manufacturer Code:NB300124
Catalog # NB300124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLUR2 glutamate metabotropic receptor 2 glutamate receptor metabotropic 2 GPRC1Bmetabotropic glutamate receptor 2 mGlu2 mGluR2 MGLUR2glutamate receptor homolog; glutamate receptor homolog; glutamate receptor, metabotropic 2; Metabotropic glutamate receptor 2; mGluR2
Gene Aliases: GLUR2; GPRC1B; GRM2; mGlu2; MGLUR2
UniProt ID: (Human) Q14416
Entrez Gene ID: (Human) 2912
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.