Invitrogen
Manufacturer Code:PA112755
Catalog # 01-674-877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the C-terminal region of mGluR5 and mGluR1as. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor family C group 1 member A; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; Metabotropic glutamate receptor 5 mGluR5 GRM5 GPRC1E MGLUR5 Metabotropic glutamate receptor 1 mGluR1 GRM1 GPRC1A MGLUR1 mGlurR1a; metabotropic glutamate receptor 5b; mGluR1; mGluR5
Gene Aliases: Gprc1a; Gprc1e; Grm1; Grm5; Mglur1; Mglur5
UniProt ID: (Rat) P23385, (Rat) P31424
Entrez Gene ID: (Rat) 24414, (Rat) 24418
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.