Invitrogen
Manufacturer Code:PA581560
Catalog # PIPA581560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Mouse GRM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member A; Metabotropic glutamate receptor 1; mGluR1; mGluR1a GPRC1A GRM1A Human metabotropic glutamate receptor 1 alpha mRNA complete cds
Gene Aliases: 4930455H15Rik; ENSMUSG00000075319; Gm10828; Gprc1a; Grm1; Mglur1; nmf373; rcw; wobl
UniProt ID: (Mouse) P97772
Entrez Gene ID: (Mouse) 14816
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.