Invitrogen
Manufacturer Code:PA14516
Catalog # APP4516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues: S(1159) S S VP S S P V S E S V L(1171) of the human GluR1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member A; glutamate receptor, metabotropic 1; Metabotropic glutamate receptor 1; mGluR1; protein phosphatase 1, regulatory subunit 85
Gene Aliases: 4930455H15Rik; ENSMUSG00000075319; Gm10828; GPRC1A; GRM1; MGLU1; MGLUR1; nmf373; PPP1R85; rcw; SCAR13; wobl
UniProt ID: (Human) Q13255, (Mouse) P97772, (Rat) P23385
Entrez Gene ID: (Human) 2911, (Mouse) 14816, (Rat) 24414
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.