Abnova
Manufacturer Code:H00004041M01
Catalog # 89-014-142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E9 |
Immunogen | LRP5 (NP_002326.1 41 a.a. ~ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant LRP5.
For Research Use Only
Protein Aliases: BMND1; EVR1; EVR4; HBM; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; Low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LR3; LRP-5; LRP-7; LRP7; OPPG; OPS; OPTA1; VBCH2
Gene Aliases: BMND1; EVR1; EVR4; HBM; LR3; LRP-5; LRP5; LRP7; OPPG; OPS; OPTA1; VBCH2
UniProt ID: (Human) O75197
Entrez Gene ID: (Human) 4041
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.