Abnova
Manufacturer Code:H00009361D01P
Catalog # 89-018-160
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | LONP1 (NP_004784.2 1 a.a. ~ 959 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human LONP1 protein.
For Research Use Only
Protein Aliases: hLON; hLON ATP-dependent protease; LON; Lon protease homolog, mitochondrial; Lon protease-like protein; LONHs; LONP; LONP1; MGC1498; Mitochondrial ATP-dependent protease Lon; mitochondrial lon protease-like protein; PIM1; PRSS15; Serine protease 15
Gene Aliases: CODASS; hLON; LON; LonHS; LONP; LONP1; PIM1; PRSS15
UniProt ID: (Human) P36776
Entrez Gene ID: (Human) 9361
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.