Novus Biologicals
Manufacturer Code:45120002
Catalog # 45-120-002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | In vivo generated recominant protein fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ce-lamin; CeLam-1; Lamin-1; LMN 1 LMN1
Gene Aliases: CELE_DY3.2; DY3.2; lam-1; lmn-1
UniProt ID: (C. elegans) Q21443
Entrez Gene ID: (C. elegans) 172687
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.