Novus Biologicals
Manufacturer Code:NBP271578AF405
Catalog # N271578A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C4 |
Immunogen | Full length recombinant protein of human katanin-p80 (NP_005877) produced in HEK293T cells. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: KAT katanin (80 kDa) katanin p80 (WD repeat containing) subunit B 1 katanin p80 (WD40-containing) subunit B 1 Katanin p80 subunit B1 katanin p80 WD40-containing subunit B1 p80 katanin; katanin (80 kDa); katanin p80 (WD repeat containing) subunit B 1; katanin p80 (WD40-containing) subunit B 1; Katanin p80 subunit B1; Katanin p80 WD40 repeat-containing subunit B1; katanin p80 WD40-containing subunit B1; KATNB1; p80 katanin
Gene Aliases: KAT; KATNB1; LIS6
UniProt ID: (Human) Q9BVA0
Entrez Gene ID: (Human) 10300
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.