Abnova
Manufacturer Code:H00003594B01
Catalog # 89-005-632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | IL12RB1 (AAH29121 1 a.a. ~ 381 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human IL12RB1 protein.
For Research Use Only
Protein Aliases: CD212; cluster of differentiation 212; IL-12 receptor beta component; IL-12 receptor subunit beta-1; IL-12R subunit beta-1; IL-12R-BETA1; IL12RB; interleukin 12 receptor, beta 1; interleukin-12 receptor beta-1 chain; Interleukin-12 receptor subunit beta-1; MGC34454
Gene Aliases: CD212; IL-12R-BETA1; IL12R; IL12RB; IL12RB1; IMD30
UniProt ID: (Human) P42701
Entrez Gene ID: (Human) 3594
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.