Abnova
Manufacturer Code:H00011213D01P
Catalog # 89-018-077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IRAK3 (NP_009130.1 1 a.a. ~ 596 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human IRAK3 protein.
For Research Use Only
Protein Aliases: ASRT5; FLJ13601; IL-1 receptor-associated kinase M; Interleukin-1 receptor-associated kinase 3; IRAK-3; IRAK-M; IRAK3; IRAKM
Gene Aliases: ASRT5; IRAK3; IRAKM
UniProt ID: (Human) Q9Y616
Entrez Gene ID: (Human) 11213
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.