Abnova
Manufacturer Code:H00003440D01P
Catalog # 89-017-759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IFNA2 (NP_000596.2 1 a.a. ~ 188 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human IFNA2 protein.
For Research Use Only
Protein Aliases: alpha-2a interferon; IFN-alpha-2; IFNA; INFA2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; Interferon alpha-2; Interferon alpha-A; interferon, alpha 2; LeIF A; MGC125764; MGC125765
Gene Aliases: IFN-alphaA; IFNA; IFNA2; IFNA2A; IFNA2B; IFNA2C; INFA2
UniProt ID: (Human) P01563
Entrez Gene ID: (Human) 3440
Molecular Function:
cytokine
interferon superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.