Novus Biologicals
Manufacturer Code:NBP152640
Catalog # NBP152640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 50 and 100 of human Eukaryotic Translation Initiation Factor 6 using the numbering given in entry NP_852133.1 (GeneID 3692). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b(2)gcn B(2)GCN homolog B4 integrin interactor CAB EIF3Ap27(BBP) eIF-6 eukaryotic translation initiation factor 3A eukaryotic translation initiation factor 6 ITGB4BPintegrin beta 4 binding protein p27 beta-4 integrin-binding protein p27BBP; B(2)GCN homolog; B4 integrin interactor; CAB; eIF-6; EIF6; eukaryotic translation initiation factor 3A; Eukaryotic translation initiation factor 6; p27 beta-4 integrin-binding protein; p27(BBP)
Gene Aliases: b(2)gcn; CAB; eIF-6; EIF3A; EIF6; ITGB4BP; OK/SW-cl.27; p27(BBP); p27BBP
UniProt ID: (Human) P56537
Entrez Gene ID: (Human) 3692
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.