Abnova
Manufacturer Code:H00003622B01
Catalog # 89-005-643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ING2 (AAH30128 1 a.a. ~ 280 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ING2 protein.
For Research Use Only
Protein Aliases: ING1L; ING1Lp; Inhibitor of growth 1-like protein; inhibitor of growth family, member 2; Inhibitor of growth protein 2; p32; p33ING2
Gene Aliases: ING1L; ING2; p33ING2
UniProt ID: (Human) Q9H160
Entrez Gene ID: (Human) 3622
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.