Invitrogen
Manufacturer Code:14592095
Catalog # 14-592-095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | CXNFT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: HEP-NOS; Hepatocyte NOS; Hepatocyte NOS hepatocytes HEP-NOS inducible nitric oxide synthase Inducible NO synthase inducible NOS iNos i-NOS Inosl MAC-NOS macrophage NOS nitric oxide synthase 2 nitric oxide synthase 2 inducible nitric oxide synthase 2 inducible macrophage nitric oxide synthase 2A (inducible hepatocytes) nitric oxide synthase inducible nitric oxide synthase macrophage nitric oxide synthase-inducible NOS NOS type II NOS type II NOS2 Nos-2 NOS2A NOS-II OTTMUSP00000000202 Peptidyl-cysteine S-nitrosylase NOS2; inducible nitric oxide synthase; Inducible NO synthase; Inducible NOS; MAC-NOS; Macrophage NOS; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; NOS type II; NOS, type II; Peptidyl-cysteine S-nitrosylase NOS2
Gene Aliases: HEP-NOS; i-NOS; INOS; Inosl; NOS; Nos-2; NOS-II; NOS2; NOS2A
UniProt ID: (Human) P35228, (Mouse) P29477, (Rat) Q06518
Entrez Gene ID: (Human) 4843, (Mouse) 18126, (Rat) 24599
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.