R&D Systems
Manufacturer Code:MAB9502
Catalog # MAB9502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D2-B2 |
Immunogen | Recombinant human iNOS Pro781-His798 Accession # P35228 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.14.13.39 Hepatocyte NOS HEP-NOS Inducible NO synthase Inducible NOS INOS nitric oxide synthase 2 inducible nitric oxide synthase 2A (inducible hepatocytes) nitric oxide synthase inducible nitric oxide synthase macrophage NOS NOS type II NOS2 NOS2A Peptidyl-cysteine S-nitrosylase NOS2; HEP-NOS; Hepatocyte NOS; Inducible NO synthase; Inducible NOS; nitric oxide synthase 2, inducible; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; NOS type II; NOS, type II; Peptidyl-cysteine S-nitrosylase NOS2
Gene Aliases: HEP-NOS; INOS; NOS; NOS2; NOS2A
UniProt ID: (Human) P35228
Entrez Gene ID: (Human) 4843
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.