Novus Biologicals
Manufacturer Code:NBP1506060.25ML
Catalog # NBP15060603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminus of mouse iNOS and nNOS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.14.13.39 Hepatocyte NOS HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2 Inducible NO synthase Inducible NOS INOS nitric oxide synthase 2 inducible nitric oxide synthase 2A (inducible hepatocytes) nitric oxide synthase inducible nitric oxide synthase macrophage NOS NOS type II NOS2ANOS type II; HEP-NOS; Hepatocyte NOS; Inducible NO synthase; Inducible NOS; nitric oxide synthase 2, inducible; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; NOS type II; NOS, type II; Peptidyl-cysteine S-nitrosylase NOS2
Gene Aliases: HEP-NOS; INOS; NOS; NOS2; NOS2A
UniProt ID: (Human) P35228
Entrez Gene ID: (Human) 4843
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.