Novus Biologicals
Manufacturer Code:NB300605
Catalog # NB300605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
In vitro Assay (IV) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | iNOS Antibody was made to a synthetic peptide corresponding to residues D(17) L K E E K D I N N N V K K T(31) of mouse iNOS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.14.13.39 Hepatocyte NOS HEP-NOS Inducible NO synthase Inducible NOS INOS nitric oxide synthase 2 inducible nitric oxide synthase 2A (inducible hepatocytes) nitric oxide synthase inducible nitric oxide synthase macrophage NOS NOS type II NOS2A Peptidyl-cysteine S-nitrosylase NOS2; HEP-NOS; Hepatocyte NOS; Inducible NO synthase; Inducible NOS; nitric oxide synthase 2, inducible; nitric oxide synthase 2A (inducible, hepatocytes); Nitric oxide synthase, inducible; nitric oxide synthase, macrophage; NOS type II; NOS, type II; Peptidyl-cysteine S-nitrosylase NOS2
Gene Aliases: HEP-NOS; INOS; NOS; NOS2; NOS2A
UniProt ID: (Human) P35228
Entrez Gene ID: (Human) 4843
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.