Invitrogen
Manufacturer Code:PA5115389
Catalog # PIPA5115389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human iASPP corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AV260376 IASPP inhibitor of apoptosis stimulating protein of p53 Inhibitor of ASPP protein NFkB interacting protein 1 NFkB-interacting protein 1 Nkip1 PPP1R13BL PPP1R13B-like protein Ppp1r13l protein iASPP protein phosphatase 1 regulatory subunit 13 like protein phosphatase 1 regulatory (inhibitor) subunit 13 like protein phosphatase 1 regulatory subunit 13 like RAI RAI4 relA-associated inhibitor retinoic acid induced 4 wa3; inhibitor of apoptosis stimulating protein of p53; Inhibitor of ASPP protein; NFkB interacting protein 1; NFkB-interacting protein 1; PPP1R13B-like protein; ppp1r13l; Protein iASPP; protein phosphatase 1, regulatory (inhibitor) subunit 13 like; protein phosphatase 1, regulatory subunit 13 like; RelA-associated inhibitor; retinoic acid induced 4
Gene Aliases: AV260376; IASPP; NKIP1; PPP1R13BL; PPP1R13L; RAI; RAI4; wa3
UniProt ID: (Human) Q8WUF5, (Mouse) Q5I1X5
Entrez Gene ID: (Human) 10848, (Mouse) 333654, (Rat) 686781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.