Abnova
Manufacturer Code:H00003062B01
Catalog # 89-005-514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | HCRTR2 (NP_001517 1 a.a. ~ 444 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human HCRTR2 protein.
For Research Use Only
Protein Aliases: hypocretin (orexin) receptor 2; Hypocretin receptor type 2; Orexin receptor type 2; orexin type-2 receptor; Ox-2-R; ox2-R; OX2R
Gene Aliases: HCRTR2; OX2R
UniProt ID: (Human) O43614
Entrez Gene ID: (Human) 3062
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.