Abnova
Manufacturer Code:H00051170B01
Catalog # 89-007-603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | DHRS8 (AAH36001.1 1 a.a. ~ 300 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human DHRS8 protein.
For Research Use Only
Protein Aliases: 17-beta-HSD 11; 17-beta-HSD XI; 17-BETA-HSD11; 17-BETA-HSDXI; 17-beta-hydroxysteroid dehydrogenase 11; 17-beta-hydroxysteroid dehydrogenase type XI; 17-beta-hydroxysteroid dehydrogenase XI; CTCL tumor antigen HD-CL-03; CTCL-associated antigen HD-CL-03; Cutaneous T-cell lymphoma-associated antigen HD-CL-03; Dehydrogenase/reductase SDR family member 8; DHRS8; Estradiol 17-beta-dehydrogenase 11; hydroxysteroid (17-beta) dehydrogenase 11; PAN1B; Retinal short-chain dehydrogenase/reductase 2; RETSDR2; SDR16C2; Short chain dehydrogenase/reductase family 16C member 2; short chain dehydrogenase/reductase family 16C, member 2
Gene Aliases: 17-BETA-HSD11; 17-BETA-HSDXI; 17BHSD11; DHRS8; HSD17B11; PAN1B; PSEC0029; RETSDR2; SDR16C2; UNQ207/PRO233
UniProt ID: (Human) Q8NBQ5
Entrez Gene ID: (Human) 51170
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.