Abnova
Manufacturer Code:H00008372B01P
Catalog # 89-016-940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | HYAL3 (AAH05896.1 1 a.a. ~ 417 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human HYAL3 protein.
For Research Use Only
Protein Aliases: Hyal-3; Hyaluronidase-3; Hyaluronoglucosaminidase-3; LUCA-3; LUCA14; LUCA3; Lung carcinoma protein 3; Minna14
Gene Aliases: HYAL-3; HYAL3; LUCA-3; LUCA3
UniProt ID: (Human) O43820
Entrez Gene ID: (Human) 8372
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.