Abnova
Manufacturer Code:H00003373D01P
Catalog # 89-017-745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HYAL1 (NP_695015.1 1 a.a. ~ 253 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human HYAL1 protein.
For Research Use Only
Protein Aliases: HYAL-1; Hyaluronidase-1; Hyaluronoglucosaminidase-1; LuCa-1; LUCA1; Lung carcinoma protein 1; MGC45987; NAT6; plasma hyaluronidase; tumor suppressor LUCA-1
Gene Aliases: HYAL-1; HYAL1; LUCA1; MPS9; NAT6
UniProt ID: (Human) Q12794
Entrez Gene ID: (Human) 3373
Molecular Function: glycosidase hydrolase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.