Abnova
Manufacturer Code:H00003211M02
Catalog # 89-000-725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F9 |
Immunogen | HOXB1 (NP_002135 1 a.a. ~ 74 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HOXB1.
For Research Use Only
Protein Aliases: Homeobox protein Hox-2I; Homeobox protein Hox-B1; Hox-2.9; HOX2; HOX2I; MGC116843; MGC116844; MGC116845
Gene Aliases: HCFP3; Hox-2.9; HOX2; HOX2I; HOXB1
UniProt ID: (Human) P14653
Entrez Gene ID: (Human) 3211
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.