Novus Biologicals
Manufacturer Code:NBP250424JF646
Catalog # NBP50424646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | N10 P2D3&asteriskG2 |
Immunogen | Ovalbumin-conjugated synthetic peptide SVKQYSGKFF (C-terminal sequence) |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: dC-stretch binding protein; dC-stretch binding protein FLJ41122 heterogeneous nuclear ribonucleoprotein K hnRNP K transformation upregulated nuclear protein Transformation up-regulated nuclear protein TUNPHNRPKCSBP; Heterogeneous nuclear ribonucleoprotein K; hnRNP K; Transformation up-regulated nuclear protein; transformation upregulated nuclear protein; TUNP
Gene Aliases: AUKS; CSBP; HNRNPK; HNRPK; TUNP
UniProt ID: (Human) P61978
Entrez Gene ID: (Human) 3190
Molecular Function:
RNA binding protein
enzyme modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.