Novus Biologicals
Manufacturer Code:NBP267656
Catalog # NBP267656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SN0652 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: C1 C2 heterogeneous nuclear ribonucleoprotein C (C1/C2) heterogeneous nuclear ribonucleoproteins C1/C2 HNRNP hnRNP C1/C2 hnRNPC HNRPC MGC104306 MGC105117 MGC117353 MGC131677 nuclear ribonucleoprotein particle C1 protein nuclear ribonucleoprotein particle C2 protein SNRPC; Heterogeneous nuclear ribonucleoproteins C1/C2; hnRNP C1/C2; nuclear ribonucleoprotein particle C1 protein; nuclear ribonucleoprotein particle C2 protein
Gene Aliases: C1; C2; HNRNP; HNRNPC; HNRPC; SNRPC
UniProt ID: (Human) P07910
Entrez Gene ID: (Human) 3183
Molecular Function:
RNA binding protein
mRNA processing factor
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.