Novus Biologicals
Manufacturer Code:NBP275526
Catalog # NBP275526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB23-43 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogeneous nuclear ribonucleoprotein U heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A) hnRNP U HNRPU p120 p120 nuclear protein pp120 SAFA SAF-AhnRNPU Scaffold attachment factor A U21.1; hnRNP U; HNRNPU antisense RNA 1; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; SAF-A; Scaffold-attachment factor A
Gene Aliases: C1orf199; hnRNP U; HNRNPU; HNRNPU-AS1; HNRPU; SAF-A; SAFA; U21.1
UniProt ID: (Human) Q00839
Entrez Gene ID: (Human) 3192
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.