Novus Biologicals
Manufacturer Code:NBP157197
Catalog # NBP157197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SYNCRIP(synaptotagmin binding cytoplasmic RNA interacting protein) The peptide sequence was selected from the middle region of SYNCRIP. Peptide sequence IEIVFAKPPDQKRKERKAQRQAAKNQMYDDYYYYGPPHMPPPTRGRGRG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ3J17.2 Glycine- and tyrosine-rich RNA-binding protein GRYRBP GRY-RBPNS1-associated protein 1 heterogeneous nuclear ribonucleoprotein Q hnRNP Q hnRNP-Q HNRPQ HNRPQ1 NSAP1FLJ31626 PP68 synaptotagmin binding cytoplasmic RNA interacting protein Synaptotagmin-binding cytoplasmic RNA-interacting protein; Glycine- and tyrosine-rich RNA-binding protein; GRY-RBP; Heterogeneous nuclear ribonucleoprotein Q; hnRNP Q; NS1-associated protein 1; synaptotagmin binding, cytoplasmic RNA interacting protein; Synaptotagmin-binding, cytoplasmic RNA-interacting protein
Gene Aliases: GRY-RBP; GRYRBP; hnRNP-Q; HNRNPQ; HNRPQ; HNRPQ1; NSAP1; PP68; SYNCRIP
UniProt ID: (Human) O60506
Entrez Gene ID: (Human) 10492
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.