Invitrogen
Manufacturer Code:MA116673
Catalog # AMPA6673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D8 |
Immunogen | M19 fusion protein containing full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CEA receptor; Heterogeneous nuclear ribonucleoprotein M; heterogeneous nuclear ribonucleoprotein M HNRNPM HNRNPM4 HNRPM hnRNA-binding protein M4 HNRNPM3 hnRNA-binding protein M3 HNRNP3 N-acetylglucosamine receptor 1 M4 protein; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; M4 protein; N-acetylglucosamine receptor 1
Gene Aliases: 2610023M21Rik; AA409009; CEAR; hnRNP M; HNRNPM; HNRNPM4; HNRPM; HNRPM4; HTGR1; mKIAA4193; NAGR1
UniProt ID: (Human) P52272, (Rat) Q62826, (Mouse) Q9D0E1
Entrez Gene ID: (Human) 4670, (Bovine) 515290, (Pig) 100514153, (Rabbit) 100356502, (Rat) 116655, (Mouse) 76936
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.