Invitrogen
Manufacturer Code:MA191607
Catalog # PIMA191607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C181 |
Immunogen | M19 fusion protein containing full length native protein (purified) (Human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CEA receptor; Heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; hnRNPM Heterogeneous nuclear ribonucleoprotein M NAGR1; M4 protein; N-acetylglucosamine receptor 1
Gene Aliases: 2610023M21Rik; AA409009; CEAR; hnRNP M; HNRNPM; HNRNPM4; HNRPM; HNRPM4; HTGR1; mKIAA4193; NAGR1
UniProt ID: (Human) P52272, (Rat) Q62826, (Mouse) Q9D0E1
Entrez Gene ID: (Human) 4670, (Bovine) 515290, (Pig) 100514153, (Rat) 116655, (Mouse) 76936
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.