Novus Biologicals
Manufacturer Code:NBP297732DL594
Catalog # NB097755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human hnRNP K fragment. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dC-stretch binding protein; dC-stretch binding protein FLJ41122 heterogeneous nuclear ribonucleoprotein K hnRNP K transformation upregulated nuclear protein Transformation up-regulated nuclear protein TUNPHNRPKCSBP; Heterogeneous nuclear ribonucleoprotein K; hnRNP K; Transformation up-regulated nuclear protein; transformation upregulated nuclear protein; TUNP
Gene Aliases: AUKS; CSBP; HNRNPK; HNRPK; TUNP
UniProt ID: (Human) P61978
Entrez Gene ID: (Human) 3190
Molecular Function: RNA binding protein enzyme modulator mRNA processing factor mRNA splicing factor nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.