Invitrogen
Manufacturer Code:PA541799
Catalog # PIPA541799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human HNRPDL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A+U-rich element RNA binding factor; AU-rich element RNA-binding factor; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); Heterogeneous nuclear ribonucleoprotein D-like; Heterogeneous nuclear ribonucleoprotein D-like HNRNP JKTBP JKTBP2 laAUF1 HNRPDL; hnRNP D-like; hnRNP DL; hnRNP-DL; hnRPD-like protein; JKT41-binding protein; Protein laAUF1
Gene Aliases: AA407431; AA959857; D5Ertd650e; D5Wsu145e; HNRNP; hnRNP DL; hnRNP-DL; HNRNPD; HNRNPDL; Hnrnpdl1; HNRPDL; JKTBP; JKTBP2; laAUF1; LGMD1G
UniProt ID: (Human) O14979, (Rat) Q3SWU3
Entrez Gene ID: (Horse) 100034002, (Human) 9987, (Dog) 476976, (Bovine) 534770, (Rat) 305178, (Mouse) 50926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.