Novus Biologicals
Manufacturer Code:NBP216863
Catalog # NBP216863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human hnRNP A2B1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: DKFZp779B0244 FLJ22720 heterogeneous nuclear ribonucleoprotein A2/B1 heterogeneous nuclear ribonucleoprotein B1 heterogeneous nuclear ribonucleoproteins A2/B1 hnRNP A2 hnRNP B1 HNRNPA2 HNRNPB1 HNRPA2 HNRPA2B1hnRNP A2/B1 HNRPB1 nuclear ribonucleoprotein particle A2 protein RNPA2 SNRPB1; Heterogeneous nuclear ribonucleoproteins A2/B1; hnRNP A2 / hnRNP B1; hnRNP A2/B1; nuclear ribonucleoprotein particle A2 protein
Gene Aliases: HNRNPA2; HNRNPA2B1; HNRNPB1; HNRPA2; HNRPA2B1; HNRPB1; IBMPFD2; RNPA2; SNRPB1
UniProt ID: (Human) P22626
Entrez Gene ID: (Human) 3181
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.