Invitrogen
Manufacturer Code:MA535132
Catalog # PIMA535132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0633 |
Immunogen | A synthesized peptide derived from human hnRNP A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ALS19 ALS20 BOS_4964 Fli-2 HDP HDP-1 Helix-destabilizing protein Heterogeneous nuclear ribonucleoprotein A1 Heterogeneous nuclear ribonucleoprotein A1 N-terminally processed hnRNP A1 hnRNP A1-like 3 hnRNP core protein A1 hnRNP I Hnrnpa1 hnrnp-A1 HNRNPI HNRNP-I Hnrpa1 HNRPA1L3 HNRPA1MGC102835 HNRPI I79_022927 IBMPFD3 MGC102835 pPTB PTB PTB-1 PTB2 PTB3 PTB4 PTB-T ROA1 RP11-78J21.1 Single-strand RNA-binding protein Single-strand-binding protein Tis Topoisomerase-inhibitor suppressed Unwinding protein 1 UP 1 UP1; HDP; HDP-1; Helix-destabilizing protein; Heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP core protein A1; hnRNP core protein A1-like 3; nuclear ribonucleoprotein particle A1 protein; putative heterogeneous nuclear ribonucleoprotein A1-like 3; single-strand DNA-binding protein UP1; Single-strand RNA-binding protein; Single-strand-binding protein; Topoisomerase-inhibitor suppressed
Gene Aliases: ALS19; ALS20; Fli-2; Hdp; HDP-1; hnRNP A1; hnRNP-A1; HNRNPA1; HNRPA1; HNRPA1L3; IBMPFD3; Tis; UP 1
UniProt ID: (Human) P09651, (Mouse) P49312, (Rat) P04256
Entrez Gene ID: (Human) 3178, (Mouse) 15382, (Rat) 29578
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.