Invitrogen
Manufacturer Code:PA579381
Catalog # PIPA579381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human HnRNP A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HDP; HDP-1; Helix-destabilizing protein; Heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1 hnRNP A1 Helix-destabilizing protein Single-strand RNA-binding protein hnRNP core protein A1 Heterogeneous nuclear ribonucleoprotein A1 N-terminally processed HNRNPA1 HNRPA1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP core protein A1; hnRNP core protein A1-like 3; nuclear ribonucleoprotein particle A1 protein; putative heterogeneous nuclear ribonucleoprotein A1-like 3; single-strand DNA-binding protein UP1; Single-strand RNA-binding protein; Single-strand-binding protein; Topoisomerase-inhibitor suppressed
Gene Aliases: ALS19; ALS20; Fli-2; Hdp; HDP-1; hnRNP A1; hnRNP-A1; HNRNPA1; HNRPA1; HNRPA1L3; IBMPFD3; Tis; UP 1
UniProt ID: (Human) P09651, (Mouse) P49312, (Rat) P04256
Entrez Gene ID: (Human) 3178, (Mouse) 15382, (Rat) 29578
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.