Invitrogen
Manufacturer Code:PA528385
Catalog # PIPA528385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 214 of Human hnRNP A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HDP; HDP-1; Helix-destabilizing protein; Heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP core protein A1; hnRNP core protein A1-like 3; HNRPA1 HNRPA1L3 hnRNP A1 hnRNP-A1 Putative heterogeneous nuclear ribonucleoprotein A1-like 3 helix-destabilizing protein heterogeneous nuclear ribonucleoprotein A1B protein heterogeneous nuclear ribonucleoprotein B2 protein heterogeneous nuclear ribonucleoprotein core protein A1 hnRNP A1-; nuclear ribonucleoprotein particle A1 protein; putative heterogeneous nuclear ribonucleoprotein A1-like 3; single-strand DNA-binding protein UP1; Single-strand RNA-binding protein; Single-strand-binding protein; Topoisomerase-inhibitor suppressed
Gene Aliases: ALS19; ALS20; Fli-2; Hdp; HDP-1; hnRNP A1; hnRNP-A1; HNRNPA1; HNRPA1; HNRPA1L3; IBMPFD3; Tis; UP 1
UniProt ID: (Human) P09651, (Mouse) P49312, (Rat) P04256
Entrez Gene ID: (Human) 3178, (Mouse) 15382, (Rat) 29578
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.