Novus Biologicals
Manufacturer Code:NBP258898
Catalog # NBP258898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GGRGGGFGGNDNFGRGGNFSGRGGFGGSRGGGGYGGSGDGYNGFGNDGGYGGGGPGYSGGSRGYGSGGQGYGNQGSGYGGSGSYDS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Helix-destabilizing protein; Helix-destabilizing protein heterogeneous nuclear ribonucleoprotein A1 heterogeneous nuclear ribonucleoprotein A1B protein heterogeneous nuclear ribonucleoprotein B2 protein heterogeneous nuclear ribonucleoprotein core protein A1 hnRNP A1 hnRNP core protein A1 hnRNPA1 hnRNP-A1 HNRPA1MGC102835 nuclear ribonucleoprotein particle A1 protein single-strand DNA-binding protein UP1 Single-strand RNA-binding protein; Heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP core protein A1; hnRNP core protein A1-like 3; nuclear ribonucleoprotein particle A1 protein; putative heterogeneous nuclear ribonucleoprotein A1-like 3; single-strand DNA-binding protein UP1; Single-strand RNA-binding protein
Gene Aliases: ALS19; ALS20; hnRNP A1; hnRNP-A1; HNRNPA1; HNRPA1; HNRPA1L3; IBMPFD3; UP 1
UniProt ID: (Human) P09651
Entrez Gene ID: (Human) 3178
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.