Abnova
Manufacturer Code:H00003208B01P
Catalog # 89-016-820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | HPCA (NP_002134 1 a.a. ~ 193 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human HPCA protein.
For Research Use Only
Protein Aliases: BDR2; Calcium-binding protein BDR-2; Neuron-specific calcium-binding protein hippocalcin
Gene Aliases: BDR2; DYT2; HPCA
UniProt ID: (Human) P84074
Entrez Gene ID: (Human) 3208
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.