Abnova
Manufacturer Code:H00008091M01
Catalog # 89-014-218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D10 |
Immunogen | HMGA2 (NP_003474 1 a.a. ~ 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HMGA2.
For Research Use Only
Protein Aliases: BABL; High mobility group AT-hook protein 2; High mobility group protein HMGI-C; high-mobility group (nonhistone chromosomal) protein isoform I-C; High-mobility group protein HMGI-C; HMGI-C; HMGIC; LIPO; STQTL9
Gene Aliases: BABL; HMGA2; HMGI-C; HMGIC; LIPO; STQTL9
UniProt ID: (Human) P52926
Entrez Gene ID: (Human) 8091
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.