Abnova
Manufacturer Code:H00003172M07
Catalog # 89-000-722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C6 |
Immunogen | HNF4A (NP_000448 324 a.a. ~ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HNF4A.
For Research Use Only
Protein Aliases: FLJ39654; hepatic nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; Hepatocyte nuclear factor 4-alpha; HNF-4-alpha; HNF4; HNF4a7; HNF4a8; HNF4a9; HNF4alpha10/11/12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor subfamily 2 group A member 1; TCF; TCF-14; TCF14; Transcription factor 14; Transcription factor HNF-4
Gene Aliases: FRTS4; HNF4; HNF4A; HNF4a7; HNF4a8; HNF4a9; HNF4alpha; MODY; MODY1; NR2A1; NR2A21; TCF; TCF14
UniProt ID: (Human) P41235
Entrez Gene ID: (Human) 3172
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.