Abnova
Manufacturer Code:H00003059M02
Catalog # 89-000-708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A8 |
Immunogen | HCLS1 (AAH16758 266 a.a. ~ 355 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HCLS1.
For Research Use Only
Protein Aliases: CTTNL; Hematopoietic cell-specific LYN substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75
Gene Aliases: CTTNL; HCLS1; HS1; lckBP1; p75
UniProt ID: (Human) P14317
Entrez Gene ID: (Human) 3059
Molecular Function: actin family cytoskeletal protein basic helix-loop-helix transcription factor cytoskeletal protein non-motor actin binding protein transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.