Abnova
Manufacturer Code:H00003070D01P
Catalog # 89-017-713
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HELLS (NP_060533.2 1 a.a. ~ 838 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human HELLS protein.
For Research Use Only
Protein Aliases: FLJ10339; LSH; Lymphoid-specific helicase; Nbla10143; PASG; Proliferation-associated SNF2-like protein; SMARCA6; SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6; SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6
Gene Aliases: HELLS; ICF4; LSH; Nbla10143; PASG; SMARCA6
UniProt ID: (Human) Q6I7N7
Entrez Gene ID: (Human) 3070
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.