Invitrogen
Manufacturer Code:PA569509
Catalog # PIPA569509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of fruit fly hb |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG9786-PA; CG9786-PB; hb CG9786; hb-PA; hb-PB; LD14196p; Protein hunchback; Regulator-of-postbithorax
Gene Aliases: CG9786; Dmel\CG9786; Dmel_CG9786; HB; hbHLH; Hunchback; l(3)85Ah; R-pbx; Rg-bx; Rg-pbx
UniProt ID: (Fruit fly) P05084
Entrez Gene ID: (Fruit fly) 41032
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.