Abnova
Manufacturer Code:PAB7333
Catalog # 89-133-156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to zebrafish hand2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of hand2.
For Research Use Only
Protein Aliases: deciduum, heart, autonomic nervous system and neural crest derivatives-expressed protein 2; dhand; hands off; heart and neural crest derivatives expressed transcript 2; MGC103415; zdHAND; zgc:103415
Gene Aliases: dhand; han; zdHAND; zgc:103415
Entrez Gene ID: (Zebrafish) 58150
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.