Novus Biologicals
Manufacturer Code:NB100174G
Catalog # NB100174G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 50000000000 |
Immunogen | Amino acids 15-374 of human p84 expressed in E. coli. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HPR1THO complex subunit 1 hTREX84 Nuclear matrix protein p84 P84 P84N5 THO complex 1 tho1; hTREX84; Nuclear matrix protein p84; p84N5; THO complex subunit 1; Tho1
Gene Aliases: HPR1; P84; P84N5; THOC1
UniProt ID: (Human) Q96FV9
Entrez Gene ID: (Human) 9984
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.