Novus Biologicals
Manufacturer Code:NBP104346AF532
Catalog # NP104346A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H3 |
Immunogen | Recombinant human TRA1 (676-803aa) purified from E. coli |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein endothelial cell (HBMEC) glycoprotein glucose regulated protein 94 kDa GP96ECGP GRP-94 GRP94gp96 homolog Heat shock protein 90 kDa beta member 1 heat shock protein 90kDa beta (Grp94) member 1 TRA1endoplasmin tumor rejection antigen (gp96) 1 Tumor rejection antigen 1 Tumor rejection antigen-1 (gp96); Endoplasmin; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; gp96 homolog; GRP-94; Heat shock protein 90 kDa beta member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; stress-inducible tumor rejection antigen gp96; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1
Gene Aliases: ECGP; GP96; GRP94; HEL-S-125m; HEL35; HSP90B1; TRA1
UniProt ID: (Human) P14625
Entrez Gene ID: (Human) 7184
Molecular Function: Hsp90 family chaperone chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.