Proteintech
Manufacturer Code:169091AP150UL
Catalog # 50-173-1251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | gp210 Fusion Protein Ag10533 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gp210 KIAA0906 Nuclear pore protein gp210 nucleoporin 210kDa Nucleoporin Nup210 NUP210 POM210; Nuclear envelope pore membrane protein POM 210; Nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; Nuclear pore protein gp210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; Nucleoporin Nup210; POM210; Pore membrane protein of 210 kDa
Gene Aliases: 9830001L10; AI836801; gp190; GP210; KIAA0906; NUP210; POM210; PSEC0245
UniProt ID: (Human) Q8TEM1, (Rat) P11654, (Mouse) Q9QY81
Entrez Gene ID: (Human) 23225, (Rat) 58958, (Mouse) 54563
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.