R&D Systems
Manufacturer Code:MAB4681
Catalog # MAB4681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 125623 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse gp130 Gln23-Glu617 (predicted) Accession # Q6PDI9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD130; CD130 antigen; CD130 CD130 antigen CDw130 DKFZp564F053 gp130 gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form IL-6 receptor subunit beta IL-6R subunit beta IL-6RB IL-6R-beta IL6ST interleukin 6 signal transducer (gp130 oncostatin M receptor) interleukin receptor beta chain interleukin-6 receptor subunit beta Interleukin-6 signal transducer Membrane glycoprotein 130 membrane glycoprotein gp130 Oncostatin-M receptor subunit alpha; CDw130; gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; IL-6 receptor subunit beta; IL-6R subunit beta; interleukin receptor beta chain; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; membrane glycoprotein gp130; Oncostatin-M receptor subunit alpha
Gene Aliases: CD130; CDW130; GP130; IL-6RB; IL6ST
UniProt ID: (Human) P40189
Entrez Gene ID: (Human) 3572
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.