Invitrogen
Manufacturer Code:PA521498
Catalog # PIPA521498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 67 and 338 of Glycerol kinase 2 (Uniprot IDNo.Q14410) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:glycerol 3-phosphotransferase 2; GK 2; GKP2 GKTA ATP:glycerol 3-phosphotransferase 2 GK 2 glycerokinase 2 glycerol kinase pseudogene 2 glycerol kinase testis specific 2 glycerol kinase testis specific 2; glycerokinase 2; Glycerol kinase 2; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; Glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m
Gene Aliases: GK2; GKP2; GKTA
UniProt ID: (Human) Q14410
Entrez Gene ID: (Human) 2712
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.