Abnova
Manufacturer Code:H00005859D01P
Catalog # 89-018-414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | QARS (NP_005042.1 1 a.a. ~ 775 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human QARS protein.
For Research Use Only
Protein Aliases: GLNRS; Glutamine--tRNA ligase; glutamine-tRNA synthetase; Glutaminyl-tRNA synthetase; PRO2195
Gene Aliases: GLNRS; MSCCA; PRO2195; QARS; QARS1
UniProt ID: (Human) P47897
Entrez Gene ID: (Human) 5859
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.