Abnova
Manufacturer Code:H00002744M01
Catalog # 89-014-117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C4 |
Immunogen | GLS (NP_055720 580 a.a. ~ 669 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GLS.
For Research Use Only
Protein Aliases: AAD20; DKFZp686O15119; FLJ10358; GLS; GLS1; glutaminase C; Glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; K-glutaminase; KIAA0838; L-glutamine amidohydrolase
Gene Aliases: AAD20; GAC; GAM; GLS; GLS1; KGA; KIAA0838
UniProt ID: (Human) O94925
Entrez Gene ID: (Human) 2744
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.